Learn More
Medchemexpress LLC α-D-glucose 1-phosphate dipotassium | 5996-14-5 | MFCD00149435 | 98.0% | 372.35 | C6H15K2O11P | 100 MG
SDP
Catalog No. 5000284986
Encompass_Preferred
α-D-Glucose 1-phosphate dipotassium is the dipotassium salt of alpha-D-glucose 1-phosphate supplied as a biochemical reagent for glycobiology and enzymatic assays. It is provided as a high-purity, water-soluble material suitable for in vitro enzyme assays, substrate studies, and pathway analysis.
  • High purity (98.0%).
  • Available in small research pack sizes (including 100 mg).
  • Molecular formula C6H15K2O11P; molecular weight 372.35.
  • Recommended long-term storage at -20°C; in solution store at -80°C for extended stability.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-84986 Supplier Medchemexpress LLC Supplier No. HYW556338100MG
α-D-Glucose 1-phosphate dipotassium is the dipotassium salt of alpha-D-glucose 1-phosphate supplied as a biochemical reagent for glycobiology and enzymatic assays. It is provided as a high-purity, water-soluble material suitable for in vitro enzyme assays, substrate studies, and pathway analysis.
  • High purity (98.0%).
  • Available in small research pack sizes (including 100 mg).
  • Molecular formula C6H15K2O11P; molecular weight 372.35.
  • Recommended long-term storage at -20°C; in solution store at -80°C for extended stability.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.