Learn More
Medchemexpress LLC α-D-Glucose 1-phosphate dipotassium | 5996-14-5 | MFCD00149435 | 98.0% | 372.35 | C6H15K2O11P | 25 MG
SDP
Catalog No. 5000284985
Encompass_Preferred
α-D-Glucose 1-phosphate dipotassium is a phosphorylated glucose substrate supplied as a solid dipotassium salt for biochemical and enzymatic research. It offers high purity and defined solubility for use in metabolic assays, enzymology, and glycobiology studies, and includes documentation such as COA and SDS for laboratory handling.
  • High purity of 98.0% for reliable experimental results.
  • Water solubility approximately 35 mg/mL with warming and sonication.
  • Solid form suitable for precise weighing and solution preparation.
  • Storage recommendations provided for stability in solution and solid state.
  • Suitable for enzymatic assays and metabolic pathway studies.
Catalog No. 50-002-84985 Supplier Medchemexpress LLC Supplier No. HYW55633825MG
α-D-Glucose 1-phosphate dipotassium is a phosphorylated glucose substrate supplied as a solid dipotassium salt for biochemical and enzymatic research. It offers high purity and defined solubility for use in metabolic assays, enzymology, and glycobiology studies, and includes documentation such as COA and SDS for laboratory handling.
  • High purity of 98.0% for reliable experimental results.
  • Water solubility approximately 35 mg/mL with warming and sonication.
  • Solid form suitable for precise weighing and solution preparation.
  • Storage recommendations provided for stability in solution and solid state.
  • Suitable for enzymatic assays and metabolic pathway studies.