Learn More
Medchemexpress LLC α-D-glucose 1-phosphate dipotassium | 5996-14-5 | MFCD00149435 | 98.0% | 372.35 g/mol | C6H15K2O11P | 10 MG
SDP
Catalog No. 5000284479
Encompass_Preferred
α-D-Glucose 1-phosphate dipotassium is the dipotassium salt of α-D-glucose 1-phosphate supplied as a high-purity biochemical reagent for in vitro enzymology and glycobiology research. It is commonly used as a substrate or reagent in assays of carbohydrate metabolism and phosphorylase or synthase enzymes.
  • High purity (98.0%).
  • Dipotassium salt form for aqueous compatibility.
  • Suitable as a substrate in enzymatic and biochemical assays.
  • Available in small research sizes (5 MG-100 MG).
  • Provided with datasheet, certificate of analysis, and safety data sheet.
  • Stable under recommended storage conditions (-20°C; in solution -80°C up to 6 months).
Catalog No. 50-002-84479 Supplier Medchemexpress LLC Supplier No. HYW55633810MG
α-D-Glucose 1-phosphate dipotassium is the dipotassium salt of α-D-glucose 1-phosphate supplied as a high-purity biochemical reagent for in vitro enzymology and glycobiology research. It is commonly used as a substrate or reagent in assays of carbohydrate metabolism and phosphorylase or synthase enzymes.
  • High purity (98.0%).
  • Dipotassium salt form for aqueous compatibility.
  • Suitable as a substrate in enzymatic and biochemical assays.
  • Available in small research sizes (5 MG-100 MG).
  • Provided with datasheet, certificate of analysis, and safety data sheet.
  • Stable under recommended storage conditions (-20°C; in solution -80°C up to 6 months).