Learn More
Medchemexpress LLC α-D-glucose 1-phosphate dipotassium | 5996-14-5 | MFCD00149435 | 98.0% | 372.35 g/mol | C6H15K2O11P | 50 MG
SDP
Catalog No. 5000284192
Encompass_Preferred
α-D-Glucose 1-phosphate dipotassium is the dipotassium salt of α-D-glucose 1-phosphate, provided for laboratory biochemical assays and glycobiology research. It serves as a stable glycosyl donor for enzymatic and analytical workflows and is supplied in small milligram pack sizes for assay flexibility.
  • Dipotassium salt with improved stability compared to the free acid.
  • Suitable as a glycosyl donor in enzymatic assays.
  • High purity (≥98.0%) for analytical applications.
  • Available in small milligram pack sizes for assay flexibility.
  • CAS 5996-14-5 and molecular weight 372.35 g/mol.
Catalog No. 50-002-84192 Supplier Medchemexpress LLC Supplier No. HYW55633850MG
α-D-Glucose 1-phosphate dipotassium is the dipotassium salt of α-D-glucose 1-phosphate, provided for laboratory biochemical assays and glycobiology research. It serves as a stable glycosyl donor for enzymatic and analytical workflows and is supplied in small milligram pack sizes for assay flexibility.
  • Dipotassium salt with improved stability compared to the free acid.
  • Suitable as a glycosyl donor in enzymatic assays.
  • High purity (≥98.0%) for analytical applications.
  • Available in small milligram pack sizes for assay flexibility.
  • CAS 5996-14-5 and molecular weight 372.35 g/mol.