Learn More
Medchemexpress LLC Α-D-Glucose-1-phosphate disodium | 56401-20-8 | 99.9% | 10 MG
SDP
Catalog No. 5000339561
Encompass_Preferred
α-D-Glucose-1-phosphate disodium serves as a foundational material for the synthesis of glucuronic acid. This compound is also recognized for its diverse applications, including its role as a cytostatic agent in cardiopathic therapy, an antibiotic, and an immunosuppressive agent. Additionally, it is utilized as an element in circulatory system therapy.
  • Starting material for glucuronic acid synthesis
  • Cytostatic compound for cardiopathic therapy
  • Antibiotic
  • Immunosuppressive agent
  • Element in circulatory system therapy
Catalog No. 50-003-39561 Supplier Medchemexpress LLC Supplier No. HY12874710MG
α-D-Glucose-1-phosphate disodium serves as a foundational material for the synthesis of glucuronic acid. This compound is also recognized for its diverse applications, including its role as a cytostatic agent in cardiopathic therapy, an antibiotic, and an immunosuppressive agent. Additionally, it is utilized as an element in circulatory system therapy.
  • Starting material for glucuronic acid synthesis
  • Cytostatic compound for cardiopathic therapy
  • Antibiotic
  • Immunosuppressive agent
  • Element in circulatory system therapy