Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC α-D-Glucose-1-phosphate disodium | 56401-20-8 | 99.9% | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHY128747R

Encompass_Preferred

α-D-Glucose-1-phosphate disodium is an endogenous metabolite that serves as a precursor for glucuronic acid synthesis. It also functions as a cytostatic compound, an antibiotic, and an immunosuppressive agent, finding use in cardiopathic and circulatory system therapies.

  • Precursor for glucuronic acid synthesis
  • Cytostatic compound for cardiopathic therapy
  • Functions as an antibiotic
  • Acts as an immunosuppressive agent
  • Component in circulatory system therapy
  • Endogenous metabolite
  • Purity: 99.9%
  • Molecular weight: 304.10
  • Formula: C6H11Na2O9P
  • CAS number: 56401-20-8
  • Appearance: White to off-white solid
  • Storage: 4°C (solid); -80°C/-20°C (solutions)

Catalog No. 50-003-41800


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.