Learn More
Medchemexpress LLC α-D-Glucose-1-phosphate disodium | 56401-20-8 | 99.9% | 25 MG
SDP
Catalog No. 5000343048
Encompass_Preferred
α-D-Glucose-1-phosphate disodium is utilized as a foundational compound in the synthesis of glucuronic acid. This substance also functions as a cytostatic agent, playing a crucial role in cardiopathic therapy, and exhibits characteristics as an antibiotic, an immunosuppressive agent, and an element involved in circulatory system therapy.
  • Starting material for glucuronic acid synthesis
  • Functions as a cytostatic compound
  • Essential for cardiopathic therapy
  • Exhibits antibiotic properties
  • Acts as an immunosuppressive agent
  • Used as a circulatory system therapy element
  • Identified as an endogenous metabolite
Catalog No. 50-003-43048 Supplier Medchemexpress LLC Supplier No. 25MGHY128747
α-D-Glucose-1-phosphate disodium is utilized as a foundational compound in the synthesis of glucuronic acid. This substance also functions as a cytostatic agent, playing a crucial role in cardiopathic therapy, and exhibits characteristics as an antibiotic, an immunosuppressive agent, and an element involved in circulatory system therapy.
  • Starting material for glucuronic acid synthesis
  • Functions as a cytostatic compound
  • Essential for cardiopathic therapy
  • Exhibits antibiotic properties
  • Acts as an immunosuppressive agent
  • Used as a circulatory system therapy element
  • Identified as an endogenous metabolite