Learn More
Medchemexpress LLC α-D-glucose-1-phosphate disodium | 56401-20-8 | MFCD00150837 | 99.9% | 304.10 g·mol⁻¹ | C6H11Na2O9P | 5 MG
SDP
Catalog No. 5000263108
Encompass_Preferred
α-D-glucose-1-phosphate disodium is the disodium salt of α-D-glucose-1-phosphate provided as a high-purity analytical standard for biochemical and analytical applications, including assay calibration, method development, and metabolite reference.
  • High purity: 99.88% (HPLC) for reliable analytical results.
  • Molecular weight 304.10 g·mol⁻¹ and formula C6H11Na2O9P for accurate reference data.
  • Suitable for assay calibration and method validation.
  • Packaged in small quantities for trace analysis and limited-sample testing.
  • Stable disodium salt with good solubility in water.
Catalog No. 50-002-63108 Supplier Medchemexpress LLC Supplier No. HY128747R5MG
α-D-glucose-1-phosphate disodium is the disodium salt of α-D-glucose-1-phosphate provided as a high-purity analytical standard for biochemical and analytical applications, including assay calibration, method development, and metabolite reference.
  • High purity: 99.88% (HPLC) for reliable analytical results.
  • Molecular weight 304.10 g·mol⁻¹ and formula C6H11Na2O9P for accurate reference data.
  • Suitable for assay calibration and method validation.
  • Packaged in small quantities for trace analysis and limited-sample testing.
  • Stable disodium salt with good solubility in water.