Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC α-D-glucose-1-phosphate (disodium hydrate) | 230954-92-4 | 98.0% | 322.11 g/mol | C6H13Na2O10P | 5 MG
SDP

Catalog No. 5000263107
Encompass_Preferred

This compound is an endogenous metabolite provided for laboratory research and synthesis. It serves as a starting material for glucuronic acid synthesis and is offered as a solid or as pre-made aqueous solutions for convenience in metabolic and biochemical studies.

  • High purity: 98.0%.
  • Available as solid and 10 mM aqueous solution.
  • Includes small pack sizes for analytical work (5 mg available).
  • Molecular weight 322.11 g/mol; formula C6H13Na2O10P.
  • Store solid at -20°C; store solution at -80°C for long-term stability.
  • SDS and datasheet available for handling and storage guidance.

Catalog No. 50-002-63107 Supplier Medchemexpress LLC Supplier No. HY128747A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This compound is an endogenous metabolite provided for laboratory research and synthesis. It serves as a starting material for glucuronic acid synthesis and is offered as a solid or as pre-made aqueous solutions for convenience in metabolic and biochemical studies.

  • High purity: 98.0%.
  • Available as solid and 10 mM aqueous solution.
  • Includes small pack sizes for analytical work (5 mg available).
  • Molecular weight 322.11 g/mol; formula C6H13Na2O10P.
  • Store solid at -20°C; store solution at -80°C for long-term stability.
  • SDS and datasheet available for handling and storage guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.