Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ D-Glucose-1-Phosphate, Disodium Salt
SCI_ED

Catalog No. S25340 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Solid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25340 1 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25340
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 56401-20-8
Molecular Formula C6H13Na2O9P
Molecular Weight (g/mol) 306.114
InChI Key AQMGMJDBUKKFOG-PKXGBZFFSA-N
Synonym alpha-d-glucose-1-phosphate disodium salt tetrahydrate
PubChem CID 131855210
IUPAC Name sodium;[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] dihydrogen phosphate
SMILES C(C1C(C(C(C(O1)OP(=O)(O)O)O)O)O)O.[Na].[Na]

Specifications

Packaging Solid
Linear Formula C6H11Na2O9P·4H2O
Quantity 1 g
Formula Weight 376.16
Physical Form Solid
Chemical Name or Material D-Glucose-1-Phosphate, Disodium Salt
Safety handling Check Alert storage code Red
ShelfLife 2 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.