Learn More
Medchemexpress LLC α-D-Glucose-1-phosphate disodium (Standard) | 56401-20-8 | 99.9% | 10 MG
SDP
Catalog No. 5000339379
Encompass_Preferred
α-D-Glucose-1-phosphate disodium (Standard) is the analytical standard of α-D-Glucose-1-phosphate disodium, intended for research and analytical applications. It serves as a starting material for glucuronic acid synthesis and finds use as a cytostatic compound, an antibiotic, an immunosuppressive agent, and a circulatory system therapy element.
  • Used as a starting material for the synthesis of glucuronic acid.
  • Can be used as a cytostatic compound essential for cardiopathic therapy.
  • Can be used as an antibiotic.
  • Can be used as an immunosuppressive agent.
  • Can be used as a circulatory system therapy element.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-003-39379 Supplier Medchemexpress LLC Supplier No. HY128747R10MG
α-D-Glucose-1-phosphate disodium (Standard) is the analytical standard of α-D-Glucose-1-phosphate disodium, intended for research and analytical applications. It serves as a starting material for glucuronic acid synthesis and finds use as a cytostatic compound, an antibiotic, an immunosuppressive agent, and a circulatory system therapy element.
  • Used as a starting material for the synthesis of glucuronic acid.
  • Can be used as a cytostatic compound essential for cardiopathic therapy.
  • Can be used as an antibiotic.
  • Can be used as an immunosuppressive agent.
  • Can be used as a circulatory system therapy element.
  • Commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.