Learn More
Medchemexpress LLC Α-D-glucose-1-phosphate disodium (standard) | 56401-20-8 | 99.9% | 100 MG
SDP
Catalog No. 5000342935
Encompass_Preferred
α-D-Glucose-1-phosphate disodium (Standard) is the analytical standard of α-D-Glucose-1-phosphate disodium. It is intended for research and analytical applications. It serves as a starting material for the synthesis of glucuronic acid and can be utilized as a cytostatic compound for cardiopathic therapy, an antibiotic, an immunosuppressive agent, and a circulatory system therapy element.
  • Functions as an analytical standard for assays.
  • Utilized in qualitative, quantitative, and methodological research experiments.
  • Compatible with techniques including HPLC, GC, and MS.
  • Can serve as a starting material for glucuronic acid synthesis.
  • Applicable as a cytostatic compound for cardiopathic therapy, an antibiotic, an immunosuppressive agent, and a circulatory system therapy element.
Catalog No. 50-003-42935 Supplier Medchemexpress LLC Supplier No. HY128747R100MG
α-D-Glucose-1-phosphate disodium (Standard) is the analytical standard of α-D-Glucose-1-phosphate disodium. It is intended for research and analytical applications. It serves as a starting material for the synthesis of glucuronic acid and can be utilized as a cytostatic compound for cardiopathic therapy, an antibiotic, an immunosuppressive agent, and a circulatory system therapy element.
  • Functions as an analytical standard for assays.
  • Utilized in qualitative, quantitative, and methodological research experiments.
  • Compatible with techniques including HPLC, GC, and MS.
  • Can serve as a starting material for glucuronic acid synthesis.
  • Applicable as a cytostatic compound for cardiopathic therapy, an antibiotic, an immunosuppressive agent, and a circulatory system therapy element.