Learn More
Cambridge Isotope Laboratories D-Glucose U-13C6, 110187-42-3, MFCD00037544, 10G
Catalog No. NC0765255
Encompass
Chemical Formula: HO*CH2(*CHOH)4*CHO, Molecular Weight: 186.11, Chemical Purity: 98%.
Catalog No. NC0765255 Supplier Cambridge Isotope Laboratories Supplier No. CLM139610
Chemical Formula: HO*CH2(*CHOH)4*CHO, Molecular Weight: 186.11, Chemical Purity: 98%.