Learn More
Medchemexpress LLC D-glucose-13C-1 | 84270-10-0 | 99.9% | 181.15 | C5(13C)H12O6 | 10 MG
SDP
Catalog No. 5000299021
Encompass_Preferred
D-Glucose-13C-1 is D-(+)-glucose labeled with 13C at the C-1 position and supplied as a high-purity isotopic tracer for metabolic and biochemical studies in preweighed laboratory quantities.
  • 13C label at the C-1 position for tracing metabolic pathways.
  • High isotopic and chemical purity (99.9%).
  • Provided in small preweighed quantities suitable for analytical experiments.
  • Molecular weight 181.15 and CAS number 84270-10-0 for unambiguous identification.
  • Stable solid form for routine laboratory storage and handling.
Catalog No. 50-002-99021 Supplier Medchemexpress LLC Supplier No. HYB0389S1110MG
D-Glucose-13C-1 is D-(+)-glucose labeled with 13C at the C-1 position and supplied as a high-purity isotopic tracer for metabolic and biochemical studies in preweighed laboratory quantities.
  • 13C label at the C-1 position for tracing metabolic pathways.
  • High isotopic and chemical purity (99.9%).
  • Provided in small preweighed quantities suitable for analytical experiments.
  • Molecular weight 181.15 and CAS number 84270-10-0 for unambiguous identification.
  • Stable solid form for routine laboratory storage and handling.