Learn More
Medchemexpress LLC D-glucose-13C | 101615-88-7 | 99.7% | 181.15 | C₅¹³CH₁₂O₆ | 10 MG
SDP
Catalog No. 5000297289
Encompass_Preferred
D-Glucose-13C is a stable isotope-labeled form of D-glucose used as a metabolic tracer for studies of glucose utilization and metabolic flux. It is suitable for mass spectrometry and NMR applications where 13C labeling enables tracing of carbon flow through biochemical pathways.
  • High isotopic labeling (13C) for metabolic tracing.
  • High purity: 99.7%.
  • Molecular weight: 181.15 g/mol.
  • Available in small research pack sizes, including 10 mg.
  • Suitable for MS and NMR-based metabolic studies.
Catalog No. 50-002-97289 Supplier Medchemexpress LLC Supplier No. HYB0389S1010MG
D-Glucose-13C is a stable isotope-labeled form of D-glucose used as a metabolic tracer for studies of glucose utilization and metabolic flux. It is suitable for mass spectrometry and NMR applications where 13C labeling enables tracing of carbon flow through biochemical pathways.
  • High isotopic labeling (13C) for metabolic tracing.
  • High purity: 99.7%.
  • Molecular weight: 181.15 g/mol.
  • Available in small research pack sizes, including 10 mg.
  • Suitable for MS and NMR-based metabolic studies.