Learn More
Medchemexpress LLC D-Glucose-13C | 101615-88-7 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0389S10
Encompass_Preferred
D-Glucose-13C is the 13C labeled D-Glucose. D-Glucose is a monosaccharide, an important carbohydrate in biology, and a carbohydrate sweetener. It is a critical component of general metabolism and serves as a critical signaling molecule related to cellular metabolic status and stress response.
  • Used as a tracer in biological studies.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development.
Catalog No. 50-004-50590