Learn More
Medchemexpress LLC D-Glucose-13C-3 | 105931-74-6 | 99.7% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0389S13100MG
Encompass_Preferred
D-Glucose-13C-3 is a 13C labeled D-Glucose. D-Glucose (Glucose) is a monosaccharide and an important carbohydrate in biology. It functions as a carbohydrate sweetener, a critical component of general metabolism, and a signaling molecule related to cellular metabolic status and stress response. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has also gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-06502