Learn More
Medchemexpress LLC D-Glucose-13C-4 | 40762-22-9 | 98.0% | 1 MG
SDP
Catalog No. 5000406653
Encompass_Preferred
D-Glucose-13C-4 is the 13C labeled form of D-Glucose. D-Glucose, a monosaccharide, is a significant carbohydrate in biology, serving as a carbohydrate sweetener and a crucial component of general metabolism. It also acts as a critical signaling molecule related to both cellular metabolic status and biotic and abiotic stress responses.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-06653 Supplier Medchemexpress LLC Supplier No. HYB0389S161MG
D-Glucose-13C-4 is the 13C labeled form of D-Glucose. D-Glucose, a monosaccharide, is a significant carbohydrate in biology, serving as a carbohydrate sweetener and a crucial component of general metabolism. It also acts as a critical signaling molecule related to both cellular metabolic status and biotic and abiotic stress responses.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.