Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-glucose-13C-4 | 40762-22-9 | 98.0% | 10 MG
SDP

Supplier:  Medchemexpress LLC HYB0389S1610MG

Encompass_Preferred

D-Glucose-13C-4 is a 13C-labeled form of D-(+)-glucose with the stable isotope at the carbon-4 position, supplied as a powder for laboratory research applications.

  • 13C label at carbon-4 for metabolic tracing and analytical studies.
  • Purity 98.0%.
  • Molecular weight 181.15 g/mol.
  • CAS number 40762-22-9.
  • Supplied as a powder; storage: powder -20°C (3 years) or 4°C (2 years).
  • Available in small packaging sizes, including 10 mg.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-98926


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.