Learn More
Medchemexpress LLC D-Glucose-13C-4 | 40762-22-9 | 98.0% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0389S16
Encompass_Preferred
D-Glucose-13C-4 is a 13C labeled D-Glucose, a monosaccharide and an important carbohydrate in biology. It serves as a critical signaling molecule related to cellular metabolic status and stress response. This compound is suitable for use as a tracer and internal standard in quantitative analyses such as NMR, GC-MS, or LC-MS.
  • 13C labeled D-Glucose
  • Important carbohydrate in biology
  • Carbohydrate sweetener
  • Critical component of general metabolism
  • Critical signaling molecule in relation to cellular metabolic status and stress response
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-50615