Learn More
Medchemexpress LLC D-Glucose-13C-4 | 40762-22-9 | 98.0% | 50 MG
SDP
Catalog No. 5000406407
Encompass_Preferred
D-Glucose-13C-4 is the 13C labeled D-Glucose. D-Glucose (Glucose), a monosaccharide, is an important carbohydrate in biology, a carbohydrate sweetener, and a critical component of general metabolism. It serves as a critical signaling molecule in relation to both cellular metabolic status and biotic and abiotic stress response. This product is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06407 Supplier Medchemexpress LLC Supplier No. HYB0389S1650MG
D-Glucose-13C-4 is the 13C labeled D-Glucose. D-Glucose (Glucose), a monosaccharide, is an important carbohydrate in biology, a carbohydrate sweetener, and a critical component of general metabolism. It serves as a critical signaling molecule in relation to both cellular metabolic status and biotic and abiotic stress response. This product is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS