Learn More
Medchemexpress LLC D-Glucose-13C-4 | 40762-22-9 | MFCD00037545 | >98.0% | 5 MG
SDP
Catalog No. 5000407319
Encompass_Preferred
D-Glucose-13C-4 is the 13C labeled D-Glucose. D-Glucose (Glucose) is a monosaccharide, an important carbohydrate in biology, a carbohydrate sweetener, and a critical component of general metabolism. It also serves as a critical signaling molecule related to both cellular metabolic status and biotic and abiotic stress response.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-07319 Supplier Medchemexpress LLC Supplier No. HYB0389S165MG
D-Glucose-13C-4 is the 13C labeled D-Glucose. D-Glucose (Glucose) is a monosaccharide, an important carbohydrate in biology, a carbohydrate sweetener, and a critical component of general metabolism. It also serves as a critical signaling molecule related to both cellular metabolic status and biotic and abiotic stress response.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.