Learn More
Medchemexpress LLC D-(+)-glucose-13C-5 | 106032-62-6 | 98.0% | 181.15 g/mol | C5(13)CH12O6 | 10 MG
SDP
Catalog No. 5000298927
Encompass_Preferred
D-Glucose-13C-5 is a stable isotope-labeled form of D-glucose with a 13C label at carbon 5, supplied as a powder for metabolic tracing and analytical applications. It is provided with defined purity, solubility, and storage conditions.
Catalog No. 50-002-98927 Supplier Medchemexpress LLC Supplier No. HYB0389S1710MG
D-Glucose-13C-5 is a stable isotope-labeled form of D-glucose with a 13C label at carbon 5, supplied as a powder for metabolic tracing and analytical applications. It is provided with defined purity, solubility, and storage conditions.