Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-Glucose-13C-5 | 106032-62-6 | >98.0% | 50 MG
SDP

Catalog No. 5000406651
Encompass_Preferred

D-Glucose-13C-5 is the 13C labeled D-Glucose. D-Glucose (Glucose), a monosaccharide, is a significant carbohydrate in biology. It serves as a carbohydrate sweetener and a critical component of general metabolism, also functioning as a crucial signaling molecule related to both cellular metabolic status and biotic and abiotic stress responses. Stable heavy isotopes, such as carbon-13, are incorporated into drug molecules for use as tracers in quantitative analysis during drug development.

  • Labeled with 13C isotope
  • Monosaccharide, significant carbohydrate in biology
  • Functions as a crucial signaling molecule
  • Used as a tracer in quantitative analysis
  • Molecular formula C5¹³CH12O6
  • Molecular weight 181.15
  • Target: Endogenous Metabolite; Isotope-Labeled Compounds
  • Pathway: Metabolic Enzyme/Protease; Others
  • In Vitro Solubility: H2O: 30 mg/mL (165.61 mM; Need ultrasonic)

Catalog No. 50-004-06651 Supplier Medchemexpress LLC Supplier No. HYB0389S1750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Glucose-13C-5 is the 13C labeled D-Glucose. D-Glucose (Glucose), a monosaccharide, is a significant carbohydrate in biology. It serves as a carbohydrate sweetener and a critical component of general metabolism, also functioning as a crucial signaling molecule related to both cellular metabolic status and biotic and abiotic stress responses. Stable heavy isotopes, such as carbon-13, are incorporated into drug molecules for use as tracers in quantitative analysis during drug development.

  • Labeled with 13C isotope
  • Monosaccharide, significant carbohydrate in biology
  • Functions as a crucial signaling molecule
  • Used as a tracer in quantitative analysis
  • Molecular formula C5¹³CH12O6
  • Molecular weight 181.15
  • Target: Endogenous Metabolite; Isotope-Labeled Compounds
  • Pathway: Metabolic Enzyme/Protease; Others
  • In Vitro Solubility: H2O: 30 mg/mL (165.61 mM; Need ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.