Learn More
Medchemexpress LLC D-glucose-13C,d-1 | 201417-06-3 | 99.7% | 182.15 | C5(13C)H11DO6 | 10 MG
SDP
Catalog No. 5000298908
Encompass_Preferred
D-Glucose-13C,d-1 is an isotopically labeled form of D-(+)-glucose used as a stable isotope tracer for metabolic and analytical studies. It is supplied as a powder with high isotopic and chemical purity for research applications.
  • Single 13C label and one deuterium substitution (d-1).
  • High chemical purity (99.7%).
  • Molecular weight approximately 182.15 g/mol.
  • Powder form with defined storage: powder -20°C (3 years) or 4°C (2 years).
  • Available in small research pack sizes, including 10 mg.
Catalog No. 50-002-98908 Supplier Medchemexpress LLC Supplier No. HYB0389S2010MG
D-Glucose-13C,d-1 is an isotopically labeled form of D-(+)-glucose used as a stable isotope tracer for metabolic and analytical studies. It is supplied as a powder with high isotopic and chemical purity for research applications.
  • Single 13C label and one deuterium substitution (d-1).
  • High chemical purity (99.7%).
  • Molecular weight approximately 182.15 g/mol.
  • Powder form with defined storage: powder -20°C (3 years) or 4°C (2 years).
  • Available in small research pack sizes, including 10 mg.