Learn More
Medchemexpress LLC D-Glucose-13C,d-1 | 201417-06-3 | 99.7% | 5 MG
SDP
Catalog No. 5000406099
Encompass_Preferred
D-Glucose-13C,d-1 is a deuterium and 13C labeled form of D-Glucose. D-Glucose (Glucose) is a monosaccharide and an important carbohydrate in biology. It functions as a carbohydrate sweetener and is a critical component of general metabolism.
  • Can be utilized as a tracer
  • Can be utilized as an internal standard for quantitative analysis
  • Useful in techniques such as NMR, GC-MS, or LC-MS
Catalog No. 50-004-06099 Supplier Medchemexpress LLC Supplier No. HYB0389S205MG
D-Glucose-13C,d-1 is a deuterium and 13C labeled form of D-Glucose. D-Glucose (Glucose) is a monosaccharide and an important carbohydrate in biology. It functions as a carbohydrate sweetener and is a critical component of general metabolism.
  • Can be utilized as a tracer
  • Can be utilized as an internal standard for quantitative analysis
  • Useful in techniques such as NMR, GC-MS, or LC-MS