Learn More
Medchemexpress LLC D-Glucose-13C,d | 83379-46-8 | 1 MG
SDP
Catalog No. 5000406095
Encompass_Preferred
D-Glucose-13C,d is the deuterium and 13C labeled D-Glucose. D-Glucose (Glucose) is a monosaccharide and an important carbohydrate in biology, serving as a critical component of general metabolism and a carbohydrate sweetener. This compound can be used as a tracer and as an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes for drug development processes
Catalog No. 50-004-06095 Supplier Medchemexpress LLC Supplier No. HYB0389S201MG
D-Glucose-13C,d is the deuterium and 13C labeled D-Glucose. D-Glucose (Glucose) is a monosaccharide and an important carbohydrate in biology, serving as a critical component of general metabolism and a carbohydrate sweetener. This compound can be used as a tracer and as an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes for drug development processes