Learn More
Medchemexpress LLC D-Glucose-13C2 | 201741-04-0 | 98.0% | 100 MG
SDP
Catalog No. 5000407318
Encompass_Preferred
D-Glucose-13C2 is the 13C labeled D-Glucose, a critical monosaccharide for general metabolism and a signaling molecule related to cellular metabolic status and stress response. This compound can be used as a tracer or an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-07318 Supplier Medchemexpress LLC Supplier No. HYB0389S14100MG
D-Glucose-13C2 is the 13C labeled D-Glucose, a critical monosaccharide for general metabolism and a signaling molecule related to cellular metabolic status and stress response. This compound can be used as a tracer or an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS