Learn More
Medchemexpress LLC D-glucose-13C2 | 201741-04-0 | 98.0% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0389S14
Encompass_Preferred
D-Glucose-13C2 is the 13C labeled D-Glucose, a monosaccharide and important carbohydrate in biology. It serves as a carbohydrate sweetener and a critical component of general metabolism, acting as a signaling molecule in relation to both cellular metabolic status and biotic and abiotic stress response.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-50558