Learn More
Medchemexpress LLC D-Glucose-13C2 | 201741-04-0 | 98.0% | 5 MG
SDP
Catalog No. 5000406652
Encompass_Preferred
D-Glucose-13C2 is a 13C labeled form of D-Glucose, a vital monosaccharide in biology. D-Glucose functions as a carbohydrate sweetener and plays a crucial role in general metabolism. It also serves as a critical signaling molecule involved in cellular metabolic status and responses to biotic and abiotic stress.
  • Can be used as a tracer for various applications.
  • Serves as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
  • Incorporation of stable heavy isotopes aids in quantitation during drug development.
Catalog No. 50-004-06652 Supplier Medchemexpress LLC Supplier No. HYB0389S145MG
D-Glucose-13C2 is a 13C labeled form of D-Glucose, a vital monosaccharide in biology. D-Glucose functions as a carbohydrate sweetener and plays a crucial role in general metabolism. It also serves as a critical signaling molecule involved in cellular metabolic status and responses to biotic and abiotic stress.
  • Can be used as a tracer for various applications.
  • Serves as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS.
  • Incorporation of stable heavy isotopes aids in quantitation during drug development.