Learn More
Medchemexpress LLC D-Glucose-13C2 | 201741-04-0 | 98.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0389S1450MG
Encompass_Preferred
D-Glucose-13C2 is a 13C labeled D-Glucose, a vital monosaccharide and carbohydrate in biology. It plays a critical role in general metabolism and functions as a signaling molecule for cellular metabolic status and stress response. This compound is valuable as a tracer and internal standard for quantitative analysis in various analytical methods.
  • 13C labeled D-Glucose.
  • Important carbohydrate and monosaccharide in biology.
  • Critical for general metabolism.
  • Serves as a signaling molecule for cellular metabolic status and stress response.
  • Can be used as a tracer in drug development.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-07124