Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC D-glucose-13C2 | 201741-04-0 | MFCD00144082 | 98.0% | 182.14 g/mol | C4(13)C2H12O6 | 10 MG
SDP

Catalog No. 5000298093
Encompass_Preferred

D-Glucose-13C2 is a 13C2-labeled form of D-glucose provided as a purified stable-isotope tracer for metabolic, analytical, and mass spectrometry applications. It is supplied as a dry, crystalline material in pre-weighed quantities for laboratory use.

  • 13C2 isotopic labeling for metabolic tracing and flux analysis.
  • Purity 98.0% suitable for analytical and MS applications.
  • Molecular weight 182.14 g/mol and formula C4(13)C2H12O6.
  • Available in small pre-weighed packages for convenient handling.
  • Supplied as a stable, dry crystalline material for reliable storage.

Catalog No. 50-002-98093 Supplier Medchemexpress LLC Supplier No. HYB0389S1410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D-Glucose-13C2 is a 13C2-labeled form of D-glucose provided as a purified stable-isotope tracer for metabolic, analytical, and mass spectrometry applications. It is supplied as a dry, crystalline material in pre-weighed quantities for laboratory use.

  • 13C2 isotopic labeling for metabolic tracing and flux analysis.
  • Purity 98.0% suitable for analytical and MS applications.
  • Molecular weight 182.14 g/mol and formula C4(13)C2H12O6.
  • Available in small pre-weighed packages for convenient handling.
  • Supplied as a stable, dry crystalline material for reliable storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.