Learn More
Medchemexpress LLC D-Glucose-13C2-4 | 138079-87-5 | 98.0% | 25 MG
SDP
Catalog No. 5000450617
Encompass_Preferred
D-Glucose-13C2-4 is the 13C labeled D-Glucose. D-Glucose (Glucose), a monosaccharide, is an important carbohydrate in biology. It is a carbohydrate sweetener and a critical component of general metabolism, serving as a critical signaling molecule in relation to both cellular metabolic status and biotic and abiotic stress response.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-50617 Supplier Medchemexpress LLC Supplier No. 25MGHYB0389S15
Add to Cart
Add to Cart
D-Glucose-13C2-4 is the 13C labeled D-Glucose. D-Glucose (Glucose), a monosaccharide, is an important carbohydrate in biology. It is a carbohydrate sweetener and a critical component of general metabolism, serving as a critical signaling molecule in relation to both cellular metabolic status and biotic and abiotic stress response.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes have been incorporated into drug molecules for quantitation during drug development.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.