Learn More
Medchemexpress LLC D-Glucose-13C6 | 110187-42-3 | 99.9% | 1 G
SDP
Catalog No. 5000406650
Encompass_Preferred
D-Glucose-13C6 is a stable isotope-labeled counterpart of D-glucose. It can be utilized as a metabolic tracer to track glucose-related synthetic catabolism, or as a synthesis ingredient, minimal media reagent, and internal standard.
  • Stable isotope-labeled counterpart of D-glucose
  • Metabolic tracer for glucose-related synthetic catabolism
  • Synthesis ingredient
  • Minimal media reagent
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06650 Supplier Medchemexpress LLC Supplier No. HYB0389A1G
D-Glucose-13C6 is a stable isotope-labeled counterpart of D-glucose. It can be utilized as a metabolic tracer to track glucose-related synthetic catabolism, or as a synthesis ingredient, minimal media reagent, and internal standard.
  • Stable isotope-labeled counterpart of D-glucose
  • Metabolic tracer for glucose-related synthetic catabolism
  • Synthesis ingredient
  • Minimal media reagent
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS