Learn More
Medchemexpress LLC D-Glucose-13C6 | 110187-42-3 | MFCD00037544 | 99.9% | 250 MG
SDP
Catalog No. 5000407047
Encompass_Preferred
D-Glucose-13C6 is a stable isotope-labeled form of D-glucose, which is a common sugar. It is utilized as a metabolic tracer to track glucose-related synthetic catabolism, or as an ingredient in synthesis, a reagent in minimal media, and an internal standard. This compound is used for research purposes only.
  • Stable isotope-labeled compound.
  • Can be used to trace glucose-related synthetic catabolism.
  • Functions as a synthesis ingredient.
  • Serves as a minimal media reagent.
  • Acts as an internal standard.
  • Used for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Reported to lightly decrease egg hatching rates, larval development, and the number of emerged adults in certain insect models.
  • Reported to prolong development time in certain insect models.
  • Reported to lightly decrease the number and period of mating for larvae labeled flies and to a greater extent for adult labeled flies.
Catalog No. 50-004-07047 Supplier Medchemexpress LLC Supplier No. 250MGHYB0389A
D-Glucose-13C6 is a stable isotope-labeled form of D-glucose, which is a common sugar. It is utilized as a metabolic tracer to track glucose-related synthetic catabolism, or as an ingredient in synthesis, a reagent in minimal media, and an internal standard. This compound is used for research purposes only.
  • Stable isotope-labeled compound.
  • Can be used to trace glucose-related synthetic catabolism.
  • Functions as a synthesis ingredient.
  • Serves as a minimal media reagent.
  • Acts as an internal standard.
  • Used for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Reported to lightly decrease egg hatching rates, larval development, and the number of emerged adults in certain insect models.
  • Reported to prolong development time in certain insect models.
  • Reported to lightly decrease the number and period of mating for larvae labeled flies and to a greater extent for adult labeled flies.