Learn More
Medchemexpress LLC D-Glucose-13C6 | 110187-42-3 | MFCD00144088 | 99.9% | 1 ML
SDP
Catalog No. 5000406093
Encompass_Preferred
D-Glucose-13C6 is a stable isotope-labeled counterpart of D-glucose. It can be used as a metabolic tracer for glucose-related synthetic catabolism or as a synthesis ingredient, minimal media reagent, and internal standard. It is for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable isotope-labeled counterpart of D-glucose
  • Traces glucose-related synthetic catabolism
  • Synthesis ingredient
  • Minimal media reagent
Catalog No. 50-004-06093 Supplier Medchemexpress LLC Supplier No. HYB0389A10MM/1ML
D-Glucose-13C6 is a stable isotope-labeled counterpart of D-glucose. It can be used as a metabolic tracer for glucose-related synthetic catabolism or as a synthesis ingredient, minimal media reagent, and internal standard. It is for research use only.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable isotope-labeled counterpart of D-glucose
  • Traces glucose-related synthetic catabolism
  • Synthesis ingredient
  • Minimal media reagent