Learn More
Medchemexpress LLC D-Glucose-13C6 | 110187-42-3 | MFCD00160408 | 99.9% | 10 G
SDP
Catalog No. 5000407104
Encompass_Preferred
D-Glucose-13C6 is a stable isotope-labeled counterpart of D-glucose. It can be used as a metabolic tracer to trace glucose-related synthetic catabolism, or as a synthesis ingredient, minimal media reagent, and internal standard.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Identified uses include laboratory chemicals and the manufacture of substances
Catalog No. 50-004-07104 Supplier Medchemexpress LLC Supplier No. HYB0389A10G
D-Glucose-13C6 is a stable isotope-labeled counterpart of D-glucose. It can be used as a metabolic tracer to trace glucose-related synthetic catabolism, or as a synthesis ingredient, minimal media reagent, and internal standard.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Identified uses include laboratory chemicals and the manufacture of substances