Learn More
Cayman Chemical D-(+)-Glucose-13C6, Purity: ≥98%, CAS: 110187-42-3, 500 mg
SDP
Catalog No. 502933975
Encompass_Preferred
An internal standard for the quantification of D-(+)-glucose by GC- or LC-MS
Catalog No. 50-293-3975 Supplier Cayman Chemical Supplier No. 26707500 MG
An internal standard for the quantification of D-(+)-glucose by GC- or LC-MS