Learn More
Medchemexpress LLC D-glucose-18O | 3343-70-2 | 99.9% | 182.16 g/mol | C6H12O6 (contains one 18O) | 10 MG
SDP
Catalog No. 5000298017
Encompass_Preferred
D-Glucose-18O is an oxygen-18-labeled form of D-glucose used as a stable isotope tracer in metabolic and biochemical research. It facilitates tracking of oxygen incorporation and exchange in enzymatic and metabolic pathways, and is supplied as a small, high-purity solid suitable for analytical applications.
  • Stable isotope-labeled glucose for tracer studies.
  • High isotopic and chemical purity for accurate mass spectrometry.
  • Suitable for metabolic flux analysis and enzymology experiments.
  • Convenient small-format solid for analytical preparation and standards.
  • CAS number 3343-70-2; molecular weight ~182.16 g/mol.
Catalog No. 50-002-98017 Supplier Medchemexpress LLC Supplier No. HYB0389S2910MG
D-Glucose-18O is an oxygen-18-labeled form of D-glucose used as a stable isotope tracer in metabolic and biochemical research. It facilitates tracking of oxygen incorporation and exchange in enzymatic and metabolic pathways, and is supplied as a small, high-purity solid suitable for analytical applications.
  • Stable isotope-labeled glucose for tracer studies.
  • High isotopic and chemical purity for accurate mass spectrometry.
  • Suitable for metabolic flux analysis and enzymology experiments.
  • Convenient small-format solid for analytical preparation and standards.
  • CAS number 3343-70-2; molecular weight ~182.16 g/mol.