Learn More
Medchemexpress LLC D-Glucose-18O | 3343-70-2 | 99.9% | 25 MG
SDP
Catalog No. 5000450613
Encompass_Preferred
D-Glucose-18O is the 18O labeled form of D-Glucose, an important monosaccharide and carbohydrate in biology. It functions as a carbohydrate sweetener and a critical component in general metabolism, serving as a key signaling molecule.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes can be incorporated into drug molecules for tracer applications.
Catalog No. 50-004-50613 Supplier Medchemexpress LLC Supplier No. 25MGHYB0389S29
D-Glucose-18O is the 18O labeled form of D-Glucose, an important monosaccharide and carbohydrate in biology. It functions as a carbohydrate sweetener and a critical component in general metabolism, serving as a key signaling molecule.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes can be incorporated into drug molecules for tracer applications.