Learn More
Medchemexpress LLC D-Glucose-18O | 3343-70-2 | 99.9% | 5 MG
SDP
Catalog No. 5000407287
Encompass_Preferred
D-Glucose-18O is the 18O labeled D-Glucose, a monosaccharide and important carbohydrate in biology. It serves as a carbohydrate sweetener and a critical component of general metabolism, functioning as a critical signaling molecule.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes can be incorporated into drug molecules for quantitation during drug development
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-07287 Supplier Medchemexpress LLC Supplier No. HYB0389S295MG
Add to Cart
Add to Cart
D-Glucose-18O is the 18O labeled D-Glucose, a monosaccharide and important carbohydrate in biology. It serves as a carbohydrate sweetener and a critical component of general metabolism, functioning as a critical signaling molecule.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Stable heavy isotopes can be incorporated into drug molecules for quantitation during drug development
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs