Learn More
Medchemexpress LLC D-Glucose, 2-amino-2-deoxy-, 6-(hydrogen sulfate) | 91674-26-9 | 97.0% | 259.23 | 10 MG
SDP
Catalog No. 5000366297
Encompass_Preferred
Glucosamine 6-sulfate (GlcN6P) is an organic cofactor that splits phosphodiester bonds. It plays a variety of catalytic roles in glmS ribozymes.
  • Appears as a white to off-white solid
  • Soluble in H2O at 250 mg/mL (964.39 mM)
  • For research use only
Catalog No. 50-003-66297 Supplier Medchemexpress LLC Supplier No. HYW00943910MG
Glucosamine 6-sulfate (GlcN6P) is an organic cofactor that splits phosphodiester bonds. It plays a variety of catalytic roles in glmS ribozymes.
  • Appears as a white to off-white solid
  • Soluble in H2O at 250 mg/mL (964.39 mM)
  • For research use only