Learn More
Medchemexpress LLC D-glucose, 4-O-α-D-glucopyranosyl-, hydrate (1:1) | 6363-53-7 | 360.31 | 10 G
SDP
Catalog No. 5000415994
Encompass_Preferred
Maltose monohydrate is a degradative product of starch and a substrate of α-Glucosidase. This compound plays a role in protecting proteins, membranes, and the photosynthetic electron transport chain under conditions of heat and freezing stress.
  • Degradative product of starch
  • Substrate of α-glucosidase
  • Protects proteins, membranes, and photosynthetic electron transport chain under heat and freezing stress
Catalog No. 50-004-15994 Supplier Medchemexpress LLC Supplier No. HYN2024A10G
Maltose monohydrate is a degradative product of starch and a substrate of α-Glucosidase. This compound plays a role in protecting proteins, membranes, and the photosynthetic electron transport chain under conditions of heat and freezing stress.
  • Degradative product of starch
  • Substrate of α-glucosidase
  • Protects proteins, membranes, and photosynthetic electron transport chain under heat and freezing stress