Learn More
Medchemexpress LLC D-Glucose, 4-O-α-D-glucopyranosyl-, hydrate (1:1) | 6363-53-7 | 696318 | 360.31 | 25 G
SDP
Catalog No. 5000389566
Encompass_Preferred
Maltose monohydrate is a degradative product of Starch and acts as a substrate for α-Glucosidase. α-Glucosidase hydrolyzes Maltose into Glucose. This compound has the ability to protect proteins, membranes, and the photosynthetic electron transport chain from heat and freezing stress. It is intended for research use only and not for sale to patients.
  • Degradative product of starch
  • Substrate of α-glucosidase
  • Hydrolyzed to glucose by α-glucosidase
  • Protects proteins under heat and freezing stress
  • Protects membranes under heat and freezing stress
  • Protects the photosynthetic electron transport chain under heat and freezing stress
Catalog No. 50-003-89566 Supplier Medchemexpress LLC Supplier No. 25GHYN2024A
Maltose monohydrate is a degradative product of Starch and acts as a substrate for α-Glucosidase. α-Glucosidase hydrolyzes Maltose into Glucose. This compound has the ability to protect proteins, membranes, and the photosynthetic electron transport chain from heat and freezing stress. It is intended for research use only and not for sale to patients.
  • Degradative product of starch
  • Substrate of α-glucosidase
  • Hydrolyzed to glucose by α-glucosidase
  • Protects proteins under heat and freezing stress
  • Protects membranes under heat and freezing stress
  • Protects the photosynthetic electron transport chain under heat and freezing stress