Learn More
Medchemexpress LLC D-Glucose | 50-99-7 | 98.0% | 10 G
SDP
Catalog No. 5000406567
Encompass_Preferred
D-Glucose is the naturally occurring and most abundant monosaccharide, essential for general metabolism and cellular signaling related to metabolic status and stress response. It is commonly used as a laboratory chemical and for the manufacture of various substances.
  • Essential for general metabolism
  • Cellular signaling molecule
  • Used in metabolic disease research
  • Applied in food research
  • Structural classification includes sweeteners and saccharides
  • Appears as a white to off-white solid
Catalog No. 50-004-06567 Supplier Medchemexpress LLC Supplier No. HYB038910G
D-Glucose is the naturally occurring and most abundant monosaccharide, essential for general metabolism and cellular signaling related to metabolic status and stress response. It is commonly used as a laboratory chemical and for the manufacture of various substances.
  • Essential for general metabolism
  • Cellular signaling molecule
  • Used in metabolic disease research
  • Applied in food research
  • Structural classification includes sweeteners and saccharides
  • Appears as a white to off-white solid