Learn More
Medchemexpress LLC D-Glucose | 50-99-7 | 98.0% | 180.16 | 500 MG
SDP
Catalog No. 5000365727
Encompass_Preferred
D-Glucose is the naturally occurring form of glucose and the most abundant monosaccharide. It is a critical component of general metabolism and serves as a vital signaling molecule in relation to both cellular metabolic status and biotic or abiotic stress response. This product is intended for laboratory chemical applications and research use only.
  • Critical component of general metabolism
  • Vital signaling molecule for cellular metabolic status and stress response
  • Increases 5-hydroxytryptamine release from human BON cells in vitro
  • Protects islet cells in alloxan-induced diabetic rats from injury and necrosis in vivo
  • Suitable for laboratory chemical applications
Catalog No. 50-003-65727 Supplier Medchemexpress LLC Supplier No. 500MGHYB0389
D-Glucose is the naturally occurring form of glucose and the most abundant monosaccharide. It is a critical component of general metabolism and serves as a vital signaling molecule in relation to both cellular metabolic status and biotic or abiotic stress response. This product is intended for laboratory chemical applications and research use only.
  • Critical component of general metabolism
  • Vital signaling molecule for cellular metabolic status and stress response
  • Increases 5-hydroxytryptamine release from human BON cells in vitro
  • Protects islet cells in alloxan-induced diabetic rats from injury and necrosis in vivo
  • Suitable for laboratory chemical applications