Learn More
Medchemexpress LLC D-Glucose | 50-99-7 | MFCD00063684 | 98.0% | 1 G
SDP
Catalog No. 5000405474
Encompass_Preferred
D-Glucose is the naturally occurring form of glucose and the most abundant monosaccharide. It is a critical component of general metabolism and serving as a significant signaling molecule related to cellular metabolic status and biotic or abiotic stress response. This biologically active stereoisomer of glucose serves as the primary energy source for living organisms.
  • Most abundant monosaccharide
  • Critical component of general metabolism
  • Signaling molecule in cellular metabolic status and stress response
  • Primary energy source for living organisms
  • Used as a standard for total sugar estimation
  • Utilized in preparing liquid media for culturing yeast cells
  • Therapeutically used in fluid and nutrient replacement
Catalog No. 50-004-05474 Supplier Medchemexpress LLC Supplier No. HYB03891G
D-Glucose is the naturally occurring form of glucose and the most abundant monosaccharide. It is a critical component of general metabolism and serving as a significant signaling molecule related to cellular metabolic status and biotic or abiotic stress response. This biologically active stereoisomer of glucose serves as the primary energy source for living organisms.
  • Most abundant monosaccharide
  • Critical component of general metabolism
  • Signaling molecule in cellular metabolic status and stress response
  • Primary energy source for living organisms
  • Used as a standard for total sugar estimation
  • Utilized in preparing liquid media for culturing yeast cells
  • Therapeutically used in fluid and nutrient replacement