Learn More
Medchemexpress LLC D-Glucose, 6-(dihydrogen phosphate), sodium salt (1:2) | 3671-99-6 | 99.9% | 1 G
SDP
Catalog No. 5000341127
Encompass_Preferred
D-Glucose 6-phosphate disodium salt is a glucose sugar phosphorylated at the hydroxy group on carbon 6. This dianion is common in cells and plays a vital role in metabolic pathways. It is for research use only.
  • Glucose sugar phosphorylated at carbon 6
  • Common in biological cells
  • Participates in biochemical pathways such as pentose phosphate pathway and glycolysis
  • Suitable for research applications
Catalog No. 50-003-41127 Supplier Medchemexpress LLC Supplier No. HY1283741G
D-Glucose 6-phosphate disodium salt is a glucose sugar phosphorylated at the hydroxy group on carbon 6. This dianion is common in cells and plays a vital role in metabolic pathways. It is for research use only.
  • Glucose sugar phosphorylated at carbon 6
  • Common in biological cells
  • Participates in biochemical pathways such as pentose phosphate pathway and glycolysis
  • Suitable for research applications