Learn More
Medchemexpress LLC D-Glucose, 6-(dihydrogen phosphate), sodium salt (1:2) | 3671-99-6 | 99.9% | 250 MG
SDP
Catalog No. 5000342084
Encompass_Preferred
D-Glucose-6-phosphate disodium salt is a glucose sugar phosphorylated at the hydroxy group on carbon 6. This dianion is very common in cells as the majority of glucose entering a cell will become phosphorylated in this way. It is for research use only.
  • Glucose sugar phosphorylated at carbon 6
  • Common endogenous metabolite in cells
  • White to off-white solid appearance
  • High purity of 99.9%
  • Suitable for various research applications
Catalog No. 50-003-42084 Supplier Medchemexpress LLC Supplier No. 250MGHY128374
D-Glucose-6-phosphate disodium salt is a glucose sugar phosphorylated at the hydroxy group on carbon 6. This dianion is very common in cells as the majority of glucose entering a cell will become phosphorylated in this way. It is for research use only.
  • Glucose sugar phosphorylated at carbon 6
  • Common endogenous metabolite in cells
  • White to off-white solid appearance
  • High purity of 99.9%
  • Suitable for various research applications