Learn More
Medchemexpress LLC D-Glucose, 6-(dihydrogen phosphate), sodium salt (1:2) | 3671-99-6 | 99.9% | 5 G
SDP
Catalog No. 5000339105
Encompass_Preferred
D-Glucose-6-phosphate disodium salt is a glucose sugar phosphorylated at the hydroxy group on carbon 6. This dianion is very common in cells, as the majority of glucose entering a cell will become phosphorylated in this way.
  • High purity of 99.9%
  • Endogenous metabolite
  • White to off-white solid appearance
  • Soluble in water
  • Used in various disease research fields, including metabolic disease
Catalog No. 50-003-39105 Supplier Medchemexpress LLC Supplier No. HY1283745G
D-Glucose-6-phosphate disodium salt is a glucose sugar phosphorylated at the hydroxy group on carbon 6. This dianion is very common in cells, as the majority of glucose entering a cell will become phosphorylated in this way.
  • High purity of 99.9%
  • Endogenous metabolite
  • White to off-white solid appearance
  • Soluble in water
  • Used in various disease research fields, including metabolic disease